Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RpLP2
- Entrez Gene ID
-
36855
- Gene Name
- Ribosomal protein LP2
- Gene Aliases
-
BcDNA:SD22208, CG4918, Dmel\CG4918, Dmel_CG4918, LP2, RLA2_DROME, RpA1, RpP1, RplP2, Rplp2, T1, Tp1, bs34h02.y1, chr2R:12101099..12101299, rpA1
- Chromosome Location
-
Chr.2R: 16586129 - 16586732 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003274
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079040.4 |
NP_523764.1 |
1 - 1 |
479 |
60 |
View Full Product Details