Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- POSH
- Entrez Gene ID
-
36990
- Gene Name
- Plenty of SH3s
- Gene Aliases
-
155116_at, BEST:GM09451, CG4909, DPOSH, Dmel\CG4909, Dmel_CG4909, GM09451, dPOSH, l(2)k15815, posh
- Chromosome Location
-
Chr.2R: 17568732 - 17572322 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0040294
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079052.4 |
NP_523776.1 |
4 - 5 |
2384 |
72 |
GenBank mRNA |
AY051984.1 |
- |
4 - 5 |
2337 |
72 |
View Full Product Details