Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dcr-2
Entrez Gene ID
36993
Gene Name
Dicer-2
Gene Aliases
CG6493, DCR-2, DCR2, DICER, DICER-2, Dcr, Dcr2, Dicer, Dicer2, Dmel\CG6493, Dmel_CG6493, cg6493, dDcr2, dcr, dcr-2, dcr2, dic2, dicer, dicer-2, dicer2, dmDcr-2
Chromosome Location
Chr.2R: 17574979 - 17581525 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034246
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299611.1 NP_001286540.1 2 - 3 250 59
NM_079054.5 NP_523778.2 2 - 3 250 59
GenBank mRNA BT128787.1 - 2 - 3 234 59

View Full Product Details