Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Smurf
Entrez Gene ID
36999
Gene Name
SMAD specific E3 ubiquitin protein ligase
Gene Aliases
CG4943, D-smurf, DSmurf, Dmel\CG4943, Dmel_CG4943, Dsmurf, LACK, Lack, SMURF, Smurf ubiquitin ligase, d-smurf, dSmurf, dSmurf1, lack, smurf
Chromosome Location
Chr.2R: 17592791 - 17604461 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029006
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274132.1 NP_001261061.1 7 - 8 3803 64
NM_079055.3 NP_523779.1 7 - 8 3803 64
GenBank mRNA AF216521.1 - 7 - 8 3780 64
AF416571.1 - 7 - 8 3780 64
AF464851.1 - 6 - 7 2899 64
BT021410.1 - 7 - 8 3780 64

View Full Product Details