Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Acox57D-p
Entrez Gene ID
37445
Gene Name
acyl-Coenzyme A oxidase at 57D proximal
Gene Aliases
Acox57Dp, CG9707, D-acox, Dmel\CG9707, Dmel_CG9707, acox
Chromosome Location
Chr.2R: 21386309 - 21389276 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034628
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299748.1 NP_001286677.1 1 - 2 192 67
NM_079078.3 NP_523802.1 1 - 2 509 67
GenBank mRNA BT133148.1 - 1 - 2 452 67

View Full Product Details