Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mRpL43
- Entrez Gene ID
-
37750
- Gene Name
- mitochondrial ribosomal protein L43
- Gene Aliases
-
BcDNA:RE35686, CG5479, CI-B8, Dmel\CG5479, Dmel_CG5479, mRPL43, mRpL-CI-B8
- Chromosome Location
-
Chr.2R: 23698105 - 23699014 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034893
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079104.3 |
NP_523828.1 |
2 - 3 |
368 |
61 |
GenBank mRNA |
AY113453.1 |
- |
2 - 3 |
365 |
61 |
View Full Product Details