Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DnaJ-60
Entrez Gene ID
37869
Gene Name
DnaJ-like-60
Gene Aliases
CG12240, CG42567, DmJ60, Dmel\CG42567, Dmel_CG42567, DnaJ60, anon-WO0172774.147
Chromosome Location
Chr.2R: 24157668 - 24159312 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0260775
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079116.4 NP_523840.1 3 - 4 609 59
NM_138048.3 NP_611892.2 3 - 4 609 59
GenBank mRNA BT128828.1 - 3 - 4 584 59

Gene Symbol
CG42568
Entrez Gene ID
8674104
Gene Name
CG42568-PA
Gene Aliases
CG12240, CG13571, Dmel\CG42568, Dmel_CG42568
Chromosome Location
Chr.2R: 24157668 - 24159312 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0260770
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169813.2 NP_001163284.1 3 - 4 605 59
NM_001169814.2 NP_001163285.1 3 - 4 609 59
GenBank mRNA BT126284.1 - 3 - 4 588 59

View Full Product Details