Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cyp9c1
- Entrez Gene ID
-
37941
- Gene Name
- Cytochrome P450-9c1
- Gene Aliases
-
9c1, CG3616, CYP9C1, Cyp9C1, Dmel\CG3616, Dmel_CG3616, P-450, P450
- Chromosome Location
-
Chr.2R: 24631063 - 24632923 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0015040
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079126.3 |
NP_523850.1 |
1 - 2 |
305 |
75 |
GenBank mRNA |
AY118661.1 |
- |
1 - 2 |
293 |
75 |
View Full Product Details