Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- gsb-n
- Entrez Gene ID
-
38004
- Gene Name
- gooseberry-neuro
- Gene Aliases
-
BSH-4, BSH4, CG2692, Dmel\CG2692, Dmel_CG2692, GSB-N, Gsb-n, Gsb-p, GsbN, Gsbn, gsb, gsb-neuro, gsb-p, gsbN, gsbn, gsbp
- Chromosome Location
-
Chr.2R: 25040383 - 25052331 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0001147
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079138.2 |
NP_523862.1 |
1 - 2 |
386 |
88 |
GenBank mRNA |
AY071593.1 |
- |
1 - 2 |
383 |
88 |
View Full Product Details