Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mth
Entrez Gene ID
38058
Gene Name
methuselah
Gene Aliases
CG6936, Dmel\CG6936, Dmel_CG6936, MTH, Mth
Chromosome Location
Chr.3L: 343378 - 346957 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0023000
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259606.2 NP_001246535.1 5 - 6 1693 69
NM_079147.4 NP_523871.1 5 - 6 1240 69
NM_167829.3 NP_728521.1 5 - 6 1240 69
GenBank mRNA AF109308.1 - 5 - 6 1201 69
BT044433.1 - 5 - 6 1192 69

View Full Product Details