Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
trh
Entrez Gene ID
38065
Gene Name
trachealess
Gene Aliases
3.1, BP1081, CG13885, CG42865, CG6883, DMU42699, Dmel\CG42865, Dmel_CG42865, HDC07747, TRH, Trh, bHLHe13, l(3)10512
Chromosome Location
Chr.3L: 366524 - 392633 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0262139
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043110.2 NP_001036575.1 1 - 2 896 67
NM_001274278.1 NP_001261207.1 1 - 2 896 67
NM_001274279.1 NP_001261208.1 1 - 2 896 67
NM_001274281.1 NP_001261210.1 1 - 2 896 67
NM_079148.3 NP_523872.2 1 - 2 896 67
GenBank mRNA BT133348.1 - 1 - 2 896 67

View Full Product Details