Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
trh
Entrez Gene ID
38065
Gene Name
trachealess
Gene Aliases
3.1, BP1081, CG13885, CG42865, CG6883, DMU42699, Dmel\CG42865, Dmel_CG42865, HDC07747, TRH, Trh, bHLHe13, l(3)10512
Chromosome Location
Chr.3L: 366524 - 392633 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0262139
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043110.2 NP_001036575.1 7 - 8 1934 68
NM_001103991.2 NP_001097461.2 10 - 11 1950 68
NM_001274278.1 NP_001261207.1 8 - 9 2021 68
NM_001274279.1 NP_001261208.1 8 - 9 2003 68
NM_001274280.1 NP_001261209.1 11 - 12 2019 68
NM_001274281.1 NP_001261210.1 7 - 8 1916 68
NM_079148.3 NP_523872.2 8 - 9 2021 68
GenBank mRNA BT133348.1 - 7 - 8 1934 68

View Full Product Details