Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
klar
Entrez Gene ID
38067
Gene Name
klarsicht
Gene Aliases
CG17046, Dmel\CG17046, Dmel_CG17046, Klar, kls, marb
Chromosome Location
Chr.3L: 434083 - 540614 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001316
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043111.3 NP_001036576.1 12 - 13 4828 59
NM_001103993.2 NP_001097463.1 13 - 14 5479 59
NM_001274282.2 NP_001261211.1 13 - 14 5415 59
NM_001274283.2 NP_001261212.1 9 - 10 4301 59
NM_001274284.1 NP_001261213.1 4 - 5 1707 59
NM_001299959.1 NP_001286888.1 9 - 10 4301 59
NM_079149.5 NP_523873.2 13 - 14 5479 59
GenBank mRNA BT010042.1 - 12 - 13 4418 59

View Full Product Details