Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
klar
Entrez Gene ID
38067
Gene Name
klarsicht
Gene Aliases
CG17046, Dmel\CG17046, Dmel_CG17046, Klar, kls, marb
Chromosome Location
Chr.3L: 434083 - 540614 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001316
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043111.3 NP_001036576.1 15 - 16 5626 82
NM_001274282.2 NP_001261211.1 16 - 17 6213 82
NM_001274283.2 NP_001261212.1 12 - 13 5099 82
NM_001274284.1 NP_001261213.1 7 - 8 2505 82
NM_079149.5 NP_523873.2 16 - 17 6277 82
GenBank mRNA BT010042.1 - 15 - 16 5216 82

View Full Product Details