Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- emc
- Entrez Gene ID
-
38091
- Gene Name
- extra macrochaetae
- Gene Aliases
-
0094/26, 0203/10, 0587/01, 0977/09, Ach, CG1007, Dm0688, Dmel\CG1007, Dmel_CG1007, Emc, bHLHb28, gov, l(3)04322, l(3)05592, l(3)61Da, l(3)j4E11, ms(3)61CD
- Chromosome Location
-
Chr.3L: 749400 - 753492 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000575
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079152.4 |
NP_523876.2 |
1 - 2 |
827 |
147 |
GenBank mRNA |
AY069405.1 |
- |
1 - 2 |
805 |
147 |
View Full Product Details