Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AlaRS-m
Entrez Gene ID
38595
Gene Name
Alanyl-tRNA synthetase, mitochondrial
Gene Aliases
Aats-ala-m, AlaS, CG4633, Dm mt AlaRS, Dmel\CG4633, Dmel_CG4633
Chromosome Location
Chr.3L: 5146098 - 5149852 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028962
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079208.3 NP_523932.2 3 - 4 585 71
GenBank mRNA AY128439.1 - 3 - 4 538 71
BT001744.1 - 3 - 4 542 71

View Full Product Details