Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
melt
Entrez Gene ID
38785
Gene Name
melted
Gene Aliases
1441/14, CG8624, Dmel\CG8624, Dmel_CG8624, Melt, Q9VS24_DROME, anon-WO03014152.10, dMelt
Chromosome Location
Chr.3L: 7134516 - 7143074 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0023001
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014572.4 NP_001014572.1 7 - 8 3064 84
NM_001274566.1 NP_001261495.1 8 - 9 3362 84
NM_079229.4 NP_523953.2 7 - 8 3082 84
GenBank mRNA BT029129.1 - 7 - 8 3056 84

View Full Product Details