Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
corn
Entrez Gene ID
38786
Gene Name
cornetto
Gene Aliases
CG32386, CG32390, CG42278, CG8621, Dmel\CG42278, Dmel_CG42278
Chromosome Location
Chr.3L: 7142991 - 7157073 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0259173
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274567.1 NP_001261496.1 2 - 3 3037 73
NM_001274568.1 NP_001261497.1 2 - 3 3037 73
NM_079230.3 NP_523954.2 2 - 3 3037 73
NM_168193.3 NP_729220.2 2 - 3 3037 73

View Full Product Details