Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tsp66A
Entrez Gene ID
38836
Gene Name
Tetraspanin 66A
Gene Aliases
CG16991, Dm.Tsp66A, Dmel\CG16991, Dmel_CG16991
Chromosome Location
Chr.3L: 7465509 - 7468808 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035786
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202134.1 NP_001189063.1 12 - 13 1694 75
NM_001274589.2 NP_001261518.1 3 - 4 477 75
NM_168216.4 NP_729259.2 3 - 4 471 75
GenBank mRNA BT081407.1 - 12 - 13 1694 75

Gene Symbol
CG42660
Entrez Gene ID
10178939
Gene Name
CG42660-PA
Gene Aliases
Dmel\CG42660, Dmel_CG42660
Chromosome Location
Chr.3L: 7465509 - 7468808 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261536
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202133.1 NP_001189062.1 12 - 13 1694 75
NM_001300039.1 NP_001286968.1 11 - 12 1760 75
GenBank mRNA BT082101.1 - 12 - 13 1694 75

Gene Symbol
CG42661
Entrez Gene ID
10178846
Gene Name
CG42661-PA
Gene Aliases
Dmel\CG42661, Dmel_CG42661
Chromosome Location
Chr.3L: 7465509 - 7468808 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261537
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202132.1 NP_001189061.1 12 - 13 1694 75
NM_001300040.1 NP_001286969.1 11 - 12 1760 75
GenBank mRNA BT126123.1 - 12 - 13 1694 75

View Full Product Details