Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gr66a
- Entrez Gene ID
-
38935
- Gene Name
- Gustatory receptor 66a
- Gene Aliases
-
CG7189, Dmel\CG7189, Dmel_CG7189, GR66A, GR66C1, GR66a, Gr66, Gr66A, Gr66C1
- Chromosome Location
-
Chr.3L: 8305336 - 8306980 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0035870
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079247.3 |
NP_523971.3 |
1 - 2 |
242 |
59 |
View Full Product Details