The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202146.1 | NP_001189075.1 | 3 - 4 | 210 | 68 |
NM_079269.3 | NP_523993.1 | 2 - 3 | 202 | 68 |