Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LanB2
Entrez Gene ID
39118
Gene Name
Laminin B2
Gene Aliases
CG3322, CT11145, DROLAMB2A, Dmel\CG3322, Dmel_CG3322, LAMB2A, LM-B2/gamma, LMG1, Lam-B2, Lmgamma1, anon-EST:fe2H3, gamma1, knod, l(3)67BDb, lamB2, laminin, lanB2
Chromosome Location
Chr.3L: 9614914 - 9623980 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0267348
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300080.1 NP_001287009.1 5 - 6 1420 85
NM_079282.2 NP_524006.1 5 - 6 1325 85
GenBank mRNA BT021394.1 - 5 - 6 1300 85

View Full Product Details