Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nrx-IV
Entrez Gene ID
39387
Gene Name
Neurexin IV
Gene Aliases
CG682, CG6827, CNTNAP2, CT21123, Dmel\CG6827, Dmel_CG6827, Dnrx, NRX, NRX IV, Nrx, Nrx IV, NrxIV, Nx IV, l(3)68Ff, l(3)rsg23, nrx, nrx IV, nrx-IV, nrxIV, rsg23
Chromosome Location
Chr.3L: 12146519 - 12156990 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0013997
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079310.3 NP_524034.2 1 - 2 443 89
NM_168491.3 NP_729787.2 1 - 2 443 89
GenBank mRNA BT120004.1 - 1 - 2 410 89

View Full Product Details