Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hml
Entrez Gene ID
39529
Gene Name
Hemolectin
Gene Aliases
CG7002, CT21553, Dmel\CG7002, Dmel_CG7002, d-hml, hml
Chromosome Location
Chr.3L: 13846054 - 13860001 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029167
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274880.1 NP_001261809.1 11 - 12 2410 51
NM_079336.3 NP_524060.2 11 - 12 2413 51

View Full Product Details