Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Or71a
- Entrez Gene ID
-
39641
- Gene Name
- Odorant receptor 71a
- Gene Aliases
-
71B.1, 71a, CG17871, DOR14, DmOr71a, Dmel\CG17871, Dmel_CG17871, OR71a, Or71aA, Or71aD, Or71c, or71a
- Chromosome Location
-
Chr.3L: 15076907 - 15078447 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0036474
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079354.4 |
NP_524078.2 |
1 - 2 |
818 |
73 |
View Full Product Details