Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mRpS31
- Entrez Gene ID
-
39749
- Gene Name
- mitochondrial ribosomal protein S31
- Gene Aliases
-
CG5904, Dmel\CG5904, Dmel_CG5904, MRP-S31, RpS31
- Chromosome Location
-
Chr.3L: 16005193 - 16006599 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0036557
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079376.3 |
NP_524100.1 |
2 - 3 |
587 |
76 |
GenBank mRNA |
AY089653.1 |
- |
2 - 3 |
583 |
76 |
View Full Product Details