Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- tra
- Entrez Gene ID
-
39849
- Gene Name
- transformer
- Gene Aliases
-
CG16724, Dmel\CG16724, Dmel_CG16724, Dmtra, TRA, Tra
- Chromosome Location
-
Chr.3L: 16590059 - 16591050 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003741
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079390.3 |
NP_524114.1 |
1 - 2 |
45 |
59 |
View Full Product Details