Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Grd
Entrez Gene ID
39984
Gene Name
Glycine receptor
Gene Aliases
CG7446, CT22917, DmGABA(GRD), Dmel\CG7446, Dmel_CG7446, GRD
Chromosome Location
Chr.3L: 17823333 - 17839767 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001134
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079407.3 NP_524131.1 5 - 6 2049 67
GenBank mRNA BT126369.1 - 5 - 6 2049 67
X78349.1 - - 1487 67

View Full Product Details