Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MESR6
Entrez Gene ID
40083
Gene Name
Misexpression suppressor of ras 6
Gene Aliases
CG6818, Dmel\CG6818, Dmel_CG6818
Chromosome Location
Chr.3L: 19092670 - 19094760 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036846
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079430.3 NP_524154.2 1 - 2 652 86
GenBank mRNA AY118919.1 - 1 - 2 622 86

View Full Product Details