The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_079439.4 | NP_524163.1 | 1 - 1 | 1146 | 170 |
GenBank mRNA | AF189278.1 | - | 1 - 1 | 1240 | 172 |