Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ko
Entrez Gene ID
40330
Gene Name
knockout
Gene Aliases
CG10573, Dmel\CG10573, Dmel_CG10573
Chromosome Location
Chr.3L: 21075581 - 21131159 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020294
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275206.1 NP_001262135.1 1 - 2 640 68
NM_079469.3 NP_524193.2 2 - 3 1088 68
GenBank mRNA BT150244.1 - 1 - 2 638 68

View Full Product Details