Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ten-m
Entrez Gene ID
40464
Gene Name
Tenascin major
Gene Aliases
2017, 5301, BcDNA:AT25108, CG11452, CG5723, CT16449, Dmel\CG5723, Dmel_CG5723, Odz, Ten-M, Ten-mc, Ten79E, Ten[m], l(3)00844, l(3)05301, l(3)05309, l(3)rJ307, l(3)rL201, l(3)rP126, odd(Oz), odz, odz/ten-m, ten(m), ten-m, ten-m/odz, ten[m]
Chromosome Location
Chr.3L: 22293044 - 22407863 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004449
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104191.3 NP_001097661.2 8 - 9 4714 57
NM_001275282.1 NP_001262211.1 8 - 9 4714 57
NM_079491.3 NP_524215.2 6 - 7 3043 57

View Full Product Details