Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nrm
Entrez Gene ID
40515
Gene Name
neuromusculin
Gene Aliases
CG32458, CG41452, CG43079, CG8779, CT16082, Dmel\CG43079, Dmel_CG43079
Chromosome Location
Chr.3L: 22998372 - 23040300 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0262509
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043156.3 NP_001036621.1 9 - 10 1772 123
NM_001259961.2 NP_001246890.1 9 - 10 1772 123
NM_001259962.2 NP_001246891.1 9 - 10 1772 123
NM_001259964.2 NP_001246893.1 9 - 10 1772 123
NM_001275303.1 NP_001262232.1 9 - 10 1772 123
NM_079494.5 NP_524218.4 9 - 10 1772 123
GenBank mRNA BT150396.1 - 9 - 10 1590 123

View Full Product Details