Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cyp9h1
- Entrez Gene ID
-
36412
- Gene Name
- CG17577-PA
- Gene Aliases
-
9h1, CG17577, Dmel\CG17577, Dmel_CG17577
- Chromosome Location
-
Chr.2R: 12840813 - 12842558 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033775
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136976.2 |
NP_610820.1 |
1 - 2 |
759 |
88 |
View Full Product Details