Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prosap
Entrez Gene ID
50225
Gene Name
CG30483-PA
Gene Aliases
BcDNA:GH23107, CG13354, CG30483, CG8122, D-ProSAP, Dmel\CG30483, Dmel_CG30483, ProSAP, Shank, anon-WO0118547.149, prosap
Chromosome Location
Chr.2R: 14060456 - 14140902 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0040752
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259386.2 NP_001246315.2 5 - 6 2165 77
NM_001299473.1 NP_001286402.1 5 - 6 2165 77
NM_001299474.1 NP_001286403.1 5 - 6 2165 77
NM_137081.5 NP_610925.3 5 - 6 2165 77
GenBank mRNA BT044467.1 - 5 - 6 1248 77

View Full Product Details