Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Shrm
Entrez Gene ID
36592
Gene Name
Shroom
Gene Aliases
CG13942, CG13944, CG34379, CG8603, Dmel\CG34379, Dmel_CG34379, Shrooom, dShrm, dmShroom
Chromosome Location
Chr.2R: 14308142 - 14351802 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0085408
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103837.2 NP_001097307.1 4 - 5 1053 60
NM_001299488.1 NP_001286417.1 4 - 5 1256 60
GenBank mRNA BT023900.1 - 4 - 5 1033 60

View Full Product Details