Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Obp50e
- Entrez Gene ID
-
36600
- Gene Name
- Odorant-binding protein 50e
- Gene Aliases
-
CG13939, DmelObp50e, Dmel\CG13939, Dmel_CG13939
- Chromosome Location
-
Chr.2R: 14374462 - 14375385 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033931
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137115.3 |
NP_610959.2 |
1 - 2 |
155 |
76 |
View Full Product Details