Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ItgaPS4
- Entrez Gene ID
-
36693
- Gene Name
- Integrin alphaPS4 subunit
- Gene Aliases
-
CG16827, CT37393, Dmel\CG16827, Dmel_CG16827, ItgalphaPS4, alpha-PS4, alpha-ps4, alphaPS4
- Chromosome Location
-
Chr.2R: 15258761 - 15262873 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034005
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137181.3 |
NP_611025.2 |
6 - 7 |
2303 |
102 |
View Full Product Details