Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unc-5
Entrez Gene ID
36703
Gene Name
CG8166-PA
Gene Aliases
AAF7419, CG8166, CG8168, CT20824, Dmel\CG8166, Dmel_CG8166, Dunc5, UNC-5, UNC5, UNC5D, Unc-5, Unc5, Unc5d, dUnc5, unc5
Chromosome Location
Chr.2R: 15340472 - 15368300 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034013
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299521.1 NP_001286450.1 9 - 10 2702 79
NM_137189.3 NP_611033.2 8 - 9 2500 79
GenBank mRNA AY058501.1 - 8 - 9 2474 79

View Full Product Details