Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- eIF2Bgamma
- Entrez Gene ID
-
36722
- Gene Name
- eukaryotic translation initiation factor 2B subunit gamma
- Gene Aliases
-
CG8190, Dmel\CG8190, Dmel_CG8190, eIF2B-gamma
- Chromosome Location
-
Chr.2R: 15542661 - 15544409 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034029
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137202.3 |
NP_611046.2 |
2 - 3 |
753 |
88 |
View Full Product Details