Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zasp52
Entrez Gene ID
36740
Gene Name
Z band alternatively spliced PDZ-motif protein 52
Gene Aliases
CG12968, CG12969, CG30084, CG8242, Dmel\CG30084, Dmel_CG30084, Z(210), ZASP52, Zasp, Zbp210, dzasp, tun, tungus, zasp, zasp52
Chromosome Location
Chr.2R: 15762565 - 15817004 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0265991
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032249.4 NP_001027420.2 5 - 6 763 71
NM_001032250.3 NP_001027421.1 5 - 6 763 71
NM_001043086.3 NP_001036551.1 5 - 6 763 71
NM_001169692.2 NP_001163163.1 5 - 6 763 71
NM_001274076.1 NP_001261005.1 5 - 6 763 71
NM_001274077.1 NP_001261006.1 5 - 6 763 71
NM_001274078.1 NP_001261007.1 5 - 6 763 71
NM_001274079.1 NP_001261008.1 5 - 6 763 71
NM_001274080.1 NP_001261009.1 5 - 6 763 71
NM_001274082.1 NP_001261011.1 5 - 6 763 71
NM_001274083.1 NP_001261012.1 5 - 6 763 71
NM_001274084.1 NP_001261013.1 5 - 6 763 71
NM_001274085.1 NP_001261014.1 5 - 6 763 71
NM_001274086.1 NP_001261015.1 5 - 6 763 71
NM_001299538.1 NP_001286467.1 5 - 6 763 71
NM_001316421.1 NP_001303350.1 5 - 6 763 71
NM_001316423.1 NP_001303352.1 5 - 6 819 71
NM_001382054.1 NP_001369093.1 5 - 6 763 71
NM_137214.5 NP_611058.4 5 - 6 763 71
NM_145757.3 NP_665700.2 5 - 6 819 71
GenBank mRNA BT001752.1 - 5 - 6 760 71
BT022171.2 - 5 - 6 743 71
BT081376.1 - 5 - 6 741 71
JN034422.1 - 5 - 6 743 71
JN034423.1 - 5 - 6 624 71
JN034424.1 - 5 - 6 759 71
JN034425.1 - 5 - 6 743 71
JN034426.1 - 5 - 6 750 71
JN034427.1 - 5 - 6 759 71
JN034428.1 - 5 - 6 731 71
JN034429.1 - 5 - 6 750 71

View Full Product Details