Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rg
Entrez Gene ID
44531
Gene Name
rugose
Gene Aliases
AKAP, Akap550, CG15464, CG15466, CG44835, CG6775, DAKAP550, Dmel\CG44835, Dmel_CG44835, fas
Chromosome Location
Chr.X: 5085759 - 5254864 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0266098
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042796.3 NP_001036261.3 7 - 8 1609 100
NM_001144685.2 NP_001138157.2 7 - 8 1921 100
NM_001144686.2 NP_001138158.2 7 - 8 1921 100
NM_001144687.2 NP_001138159.2 7 - 8 1921 100
NM_001297949.1 NP_001284878.1 10 - 11 5938 100
NM_001297950.1 NP_001284879.1 7 - 8 1921 100
NM_167028.3 NP_726978.2 8 - 9 3282 100

Gene Symbol
CG15465
Entrez Gene ID
31434
Gene Name
CG15465-PA
Gene Aliases
Dmel\CG15465, Dmel_CG15465
Chromosome Location
Chr.X: 5085759 - 5254864 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029746
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297948.1 NP_001284877.1 10 - 11 5938 100
NM_131978.3 NP_572206.1 8 - 9 3282 100

View Full Product Details