Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Trxr-1
- Entrez Gene ID
-
31760
- Gene Name
- Thioredoxin reductase-1
- Gene Aliases
-
2151, CG2151, DTR, Dm-TrxR, DmTR, DmTrx, DmTrxR, DmTrxR-1, Dmel\CG2151, Dmel_CG2151, GR, Gr, TH, TRR, TRXR1, Trx, TrxR, TrxR-1, TrxR1, Trxr, Trxr1, anon-WO03040301.185, anon-WO03040301.187, dTrxR, dmtrxr-1, gr, l(1)G0154, l(1)G0379, l(1)G0477, l(1)G0481, trxr-1, trxr1
- Chromosome Location
-
Chr.X: 8242528 - 8248033 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0020653
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_167149.2 |
NP_727251.1 |
1 - 2 |
474 |
65 |
View Full Product Details