Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ppt1
Entrez Gene ID
31805
Gene Name
Palmitoyl-protein thioesterase 1
Gene Aliases
CG12108, DmPpt1, Dmel\CG12108, Dmel_CG12108, PPT1, ppt1
Chromosome Location
Chr.X: 8688627 - 8690227 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030057
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298095.2 NP_001285024.1 2 - 3 733 89
NM_167166.4 NP_727284.1 1 - 2 612 89
GenBank mRNA AF513720.1 - 1 - 2 554 89
BT150202.1 - 1 - 2 610 89

View Full Product Details