Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kap3
Entrez Gene ID
32071
Gene Name
Kinesin associated protein 3
Gene Aliases
CG11759, DmKAP, DmKap, Dmel\CG11759, Dmel_CG11759, KAP, KAP3, Kap, dKAP3, kap3
Chromosome Location
Chr.X: 11334187 - 11340084 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028421
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103480.2 NP_001096950.2 5 - 6 1138 73
NM_001144714.1 NP_001138186.1 3 - 4 629 73
NM_001272516.2 NP_001259445.2 4 - 5 969 73
NM_167278.3 NP_727512.3 4 - 5 969 73
GenBank mRNA BT010070.1 - 4 - 5 936 73

View Full Product Details