Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Gr10a
- Entrez Gene ID
-
117501
- Gene Name
- Gustatory receptor 10a
- Gene Aliases
-
CG32664, Dmel\CG32664, Dmel_CG32664, GR10a, Gr10B1, gr10a
- Chromosome Location
-
Chr.X: 11413467 - 11417422 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0045502
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_167283.3 |
NP_727523.1 |
5 - 6 |
2629 |
64 |
GenBank mRNA |
DQ989010.1 |
- |
1 - 2 |
1070 |
64 |
- Gene Symbol
- Or10a
- Entrez Gene ID
-
32086
- Gene Name
- Odorant receptor 10a
- Gene Aliases
-
10B.1, 10a, CG17867, DOR92, Dmel\CG17867, Dmel_CG17867, Gr10a, OR10a
- Chromosome Location
-
Chr.X: 11413467 - 11417422 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0030298
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078567.5 |
NP_511122.1 |
5 - 6 |
2629 |
64 |
View Full Product Details