Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
zfh2
Entrez Gene ID
43795
Gene Name
Zn finger homeodomain 2
Gene Aliases
CG1449, Dmel\CG1449, Dmel_CG1449, ZFH-2, ZFH2, Zfh-2, Zfh2, zfh-2
Chromosome Location
Chr.4: 501810 - 540874 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004607
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272130.1 NP_001259059.1 7 - 8 5211 75
NM_079884.5 NP_524623.2 7 - 8 5211 75

View Full Product Details