Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
apolpp
Entrez Gene ID
43827
Gene Name
apolipophorin
Gene Aliases
ApoL1, ApoL2, ApoLI, ApoLII, ApoLpp, Apolpp, CG11064, DRBP, Dmel\CG11064, Dmel_CG11064, Lipophorin, Lpp, RFABG, RFABP, RFBP, RfaBP, RfaBp, Rfabg, Rfabp, T3, apoLI, apoLII, apoLPP, apoLpp, chr4:1088013..1088173, lipophorin, rfabg
Chromosome Location
Chr.4: 1060969 - 1077273 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0087002
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272156.1 NP_001259085.1 5 - 6 7630 125
NM_001297791.1 NP_001284720.1 5 - 6 7681 125
NM_001297792.1 NP_001284721.1 5 - 6 7755 125
NM_079895.3 NP_524634.2 5 - 6 7630 125

View Full Product Details