Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
toy
Entrez Gene ID
43833
Gene Name
twin of eyeless
Gene Aliases
CG11186, Dmel\CG11186, Dmel_CG11186, Pax-6, TOY, Toy, l(4)102CDg, l(4)8
Chromosome Location
Chr.4: 989725 - 1007922 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0019650
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272151.1 NP_001259080.1 4 - 5 1367 98
NM_001382011.1 NP_001368990.1 4 - 5 1367 98
NM_079899.4 NP_524638.3 4 - 5 1367 98
GenBank mRNA AY051487.1 - 4 - 5 1339 98

View Full Product Details