Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mGluR
Entrez Gene ID
43838
Gene Name
metabotropic Glutamate Receptor
Gene Aliases
CAA67993.1_Dr, CG11144, DmGlu-A, DmGlu-RA, DmGluA, DmGluAR, DmGluR, DmGluRA, DmGluR[[A]], DmGlu[[A]]R, Dmel\CG11144, Dmel_CG11144, FBgn0019985, Glu-RA, GluR, dmGluRA, mGLuR, mGluRA, mGlurA
Chromosome Location
Chr.4: 936819 - 949441 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0019985
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272147.1 NP_001259076.1 4 - 5 1699 111
NM_001272148.1 NP_001259077.1 4 - 5 1699 111
NM_001297789.1 NP_001284718.1 3 - 4 1155 111
NM_079900.3 NP_524639.2 4 - 5 1699 111
GenBank mRNA BT088899.1 - 4 - 5 1673 111

View Full Product Details